Key Documents
D7567
Sigma-Aldrich
2′,5′-dd-3′-AMP-bis(t-Bu-SATE)
≥98% (HPLC), liquid (thick)
Synonym(s):
2′,5′-Dideoxyadenosine-3′-O-bis(S-pivaloyl-2-thioethyl)-phosphate
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C24H38N5O7PS2
CAS Number:
Molecular Weight:
603.69
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
assay
≥98% (HPLC)
form
liquid (thick)
color
clear
solubility
DMSO: 5 mg/mL
storage temp.
−20°C
SMILES string
C[C@H]1O[C@H](C[C@@H]1OP(=O)(OCCSC(=O)C(C)(C)C)OCCSC(=O)C(C)(C)C)n2cnc3c(N)ncnc23
InChI
1S/C24H38N5O7PS2/c1-15-16(12-17(35-15)29-14-28-18-19(25)26-13-27-20(18)29)36-37(32,33-8-10-38-21(30)23(2,3)4)34-9-11-39-22(31)24(5,6)7/h13-17H,8-12H2,1-7H3,(H2,25,26,27)/t15-,16+,17-/m1/s1
InChI key
LUNRTVLIFYTBNX-IXDOHACOSA-N
Biochem/physiol Actions
2′,5′-dd-3′-AMP-bis(t-Bu-SATE) is a potent, cell-permeable adenylyl cyclase inhibitor. IC50 = 30 nM.9-substituted adenine derivatives with protected phosphoryl groups were synthesized and tested as inhibitors of adenylyl cyclase in isolated enzyme and intact cell systems. Protected 3′-phosphoryl derivatives of 2′,5′-dideoxyadenosine (2′,5′-dd-Ado) and beta-l-2′,5′-dd-Ado, protected 5′-phosphoryl derivatives of beta-l-2′,3′-dd-Ado, and protected phosphoryl derivatives of two 9-(2-phosphonomethoxy-acyl)-adenines were synthesized. Protection was afforded by two cyclosaligenyl- or three S-acyl-2-thioethyl-substituents. These pro-nucleotides were tested for their capacity to block forskolin-induced increases in [3H]cAMP in OB1771 and F442A preadipocytes and human macrophages prelabeled with [3H]adenine. A striking selectivity for 2′,5′-dd-Ado-3′-phosphoryl derivatives was observed. Cyclosaligenyl-derivatives (IC50 approximately 2 microm) were much less potent than S-acyl-2-thioethyl-derivatives. Best studied of these was 2′,5′-dd-Ado-3′-O-bis(S-pivaloyl-2-thioethyl)-phosphate, which blocked [3H]cAMP formation in preadipocytes (IC50 approximately 30 nm) and suppressed opening of cAMP-dependent Cl(-) channels in cardiac myocytes (IC50 approximately 800 nm). The availability and characteristics of these prodrugs should make them useful for blocking cAMP-mediated pathways in intact cell systems, in biochemical, pharmacological, and potentially therapeutic contexts.
2′,5′-dd-3′-AMP-bis(t-Bu-SATE) is a potent, cell-permeable adenylyl cyclase inhibitor. IC50 = 30 nM.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)